Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • gamma Catenin Antibodies

          Progen

          Plakoglobin Monoclonal Antibody (PG 5.1)

          View all (46) gamma Catenin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Plakoglobin Monoclonal Antibody (PG 5.1)

          Product Details

          61005PROGEN

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:25-1:100
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:25-1:100
          -

          Immunocytochemistry (ICC/IF)

          1:5
          -
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Rat, Zebrafish

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          PG 5.1

          Immunogen

          Plakoglobin,
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 0.5% BSA

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute lyophilized product with 1 mL distilled water.

          PG 5.1 represents an excellent marker for all forms of intercellular adhering junctions, such as: desmosomes of epithelial and myocardial cells (incl. cultured cells); zonulae and fasciae adherentes of epithelia, endothelia of blood vessels and myocardial cells; adherens-type junctions (e.g. lens tissue, pigmented retinal cells, Sertoli cells of testis). The PG 5.1 epitope maps within the C-terminus at the extreme end of repeat 13 (aa 632-687) of plakoglobin. Polypeptide reacting: MW 83 000 Plakoglobin, "band 5" polypeptide of intercellular adhering junctions (identical to gamma-catenin)Reactivity on cultured cell lines: Cell cultures forming monolayers .

          Target Information

          JUP is a major cytoplasmic protein which is the only known constituent common to sub membranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: catenin (cadherin-associated protein), gamma 80kDa; Catenin gamma; Desmoplakin III; Desmoplakin-3; gama-catenin; Gamma-catenin; gamma-catenin (plakoglobin); Junction plakoglobin; Junction plakoglobin a; OTTHUMP00000164735; Plakoglobin; Plakoglobin-1a

          View more View less

          Gene Aliases: ARVD12; BOS_19078; cb80; CTNNG; D930025P04Rik; danPG; DP3; DPIII; fb11b06; JUP; jupa; PDGB; PG; PKGB; wu:fb11b06

          View more View less

          UniProt ID: (Bovine) Q8SPJ1, (Human) P14923, (Rat) Q6P0K8, (Mouse) Q02257, (Zebrafish) Q9PVF7

          View more View less

          Entrez Gene ID: (Bovine) 445543, (Human) 3728, (Rat) 81679, (Mouse) 16480, (Zebrafish) 30415

          View more View less

          Function(s)
          transcription coactivator activity structural molecule activity protein binding protein kinase binding protein phosphatase binding protein homodimerization activity alpha-catenin binding cadherin binding signal transducer activity structural constituent of cell wall cell adhesion molecule binding cell adhesive protein binding involved in bundle of His cell-Purkinje myocyte communication cadherin binding involved in cell-cell adhesion nuclear hormone receptor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter cell morphogenesis desmosome assembly negative regulation of heart induction by canonical Wnt signaling pathway atrioventricular valve morphogenesis negative regulation of Wnt signaling pathway involved in heart development cytoskeletal anchoring at plasma membrane gastrulation ectoderm development nervous system development morphogenesis of embryonic epithelium cell migration regulation of cell proliferation positive regulation of protein import into nucleus regulation of apoptotic process skin development oocyte development detection of mechanical stimulus positive regulation of sequence-specific DNA binding transcription factor activity endothelial cell-cell adhesion cellular response to indole-3-methanol ventricular cardiac muscle cell action potential bundle of His cell to Purkinje myocyte communication regulation of heart rate by cardiac conduction cell adhesion heart development negative regulation of Wnt signaling pathway positive regulation of transcription from RNA polymerase II promoter signal transduction single organismal cell-cell adhesion adherens junction organization adherens junction assembly protein heterooligomerization bundle of His cell-Purkinje myocyte adhesion involved in cell communication establishment of protein localization to plasma membrane positive regulation of canonical Wnt signaling pathway regulation of ventricular cardiac muscle cell action potential regulation of cell fate specification regulation of nucleic acid-templated transcription
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0