Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Progen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Specificity: plakophilin 1 in diverse stratified epithelia, in complex epithelia (e.g. urothelium of the bladder and tracheal epithelium) and in glandular duct epithelia; squamous cell carcinoma are positive, whereas all adenocarcinoma tested so far are negative. Polypeptide reacting: Plakophilin 1: calculated mol. wt. 80,496 (human) and 80,180 (bovine).Reactivity on human cultured cell lines: HaCaT, A-431. This antibody was used in a Indirect ELISA.
This gene encodes a member of the arm-repeat (armadillo) and plakophilin gene families. Plakophilin proteins contain numerous armadillo repeats, localize to cell desmosomes and nuclei, and participate in linking cadherins to intermediate filaments in the cytoskeleton. This protein may be involved in molecular recruitment and stabilization during desmosome formation. Mutations in this gene have been associated with the ectodermal dysplasia/skin fragility syndrome. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2010].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: armadillo; B6P; band 6; Band 6 protein; Band-6 protein; MGC138829; Plakophilin-1
Gene Aliases: B6P; BOS_16110; PKP1
UniProt ID: (Human) Q13835, (Bovine) Q28161
Entrez Gene ID: (Human) 5317, (Bovine) 281405
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support