Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: RGIKHRFKW NRKVLPSSGG QITEARVAEN RPGAFIKQGR KLDIDFGAEG NRYYAANYWQ FPDGIYYEGC SEANVTKEML VTSCVNATQA ANQAEFSREK QDSKLHQRVL WRLIKEICSA KHCDFWLER (27-154 aa encoded by BC025140)
This gene is found on chromosome 20, approximately 20 kbp downstream of the gene encoding cellular prion protein, to which it is biochemically and structurally similar. The protein encoded by this gene is a membrane glycosylphosphatidylinositol-anchored glycoprotein that is found predominantly in testis. Mutations in this gene may lead to neurological disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Doppelganger; Dpl; prion gene complex, downstream; Prion protein 2; prion protein-like protein; Prion-like protein doppel; PrPLP
Gene Aliases: AI450264; dJ1068H6.4; DOPPEL; DPL; PRND; PrPLP; UNQ1830/PRO3443
UniProt ID: (Human) Q9UKY0, (Mouse) Q9QUG3
Entrez Gene ID: (Human) 23627, (Mouse) 26434
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support