Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Protocadherin Gamma Antibodies

          Invitrogen

          Protocadherin Gamma Monoclonal Antibody (N159/5)

          View all (6) Protocadherin Gamma antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Protocadherin Gamma Monoclonal Antibody (N159/5)

          • Antibody Testing Data (2)
          Protocadherin Gamma Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Protocadherin Gamma Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Protocadherin Gamma Antibody (MA5-27615) in ICC/IF

          Immunofluorescent analysis of Protocadherin Gamma in human neuroblastoma cell line (SK-N-BE). Sample was fixed with 4% formaldehyde (15 min at RT), incubated with Protocadherin Gamma monoclonal antibody (Product # MA5-27615) using a dilution of 1:100 (1 hour at RT), and followed by Goat Anti-Mouse 488, Phalloidin Texas Red and DAPI secondary antibody at a dilution of 1:200, 1:1000 (60 min at RT) and 1:5000 ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Protocadherin Gamma Antibody in Immunocytochemistry (ICC/IF)
          Protocadherin Gamma Antibody in Western Blot (WB)

          Product Details

          MA5-27615

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1:100
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N159/5

          Immunogen

          Fusion protein corresponding to amino acids 808-931 (C-terminal cytoplasmic constant domain) of mouse Protocadherin-gamma-A1 that is shared by all 22 Gamma-protocadherins (A subfamily amino acids ~807-930, B subfamily amino acids ~789-912 and C subfamily amino acids ~818-941). 99% identity with human (123/124 amino acids).

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • APC 
          • FITC
          • PE 
          • PerCP 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2735332

          Product Specific Information

          1 µg/mL of MA5-27615 was sufficient for detection of Protocadherin gamma (pan) in 20 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 100kDa. Cross-reacts with all Gamma-protocadherin-A, -B and -C proteins.


          This antibody was formerly sold as clone S159-5.

          Target Information

          The protocadherin gamma gene cluster is one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 3' RACE product; cadherin 20; cadherin 21; cadherin ME3; cadherin ME6; Cadherin-20; Cadherin-21; cadherin-like 2; fibroblast cadherin FIB2; fibroblast cadherin FIB3; Fibroblast cadherin-2; Fibroblast cadherin-3; Pan-Gamma-Protocadherin-A; PC-43; PCDH gamma A; PCDH-gamma-A; PCDH-gamma-A1; PCDH-gamma-A10; PCDH-gamma-A11; PCDH-gamma-A12; PCDH-gamma-A2; PCDH-gamma-A3; PCDH-gamma-A4; PCDH-gamma-A5; PCDH-gamma-A6; PCDH-gamma-A7; PCDH-gamma-A8; PCDH-gamma-A9; PCDH-gamma-B1; PCDH-gamma-B2; PCDH-gamma-B3; PCDH-gamma-B4; PCDH-gamma-B5; PCDH-gamma-B6; PCDH-gamma-B7; PCDH-gamma-C3; PCDH-gamma-C4; PCDH-gamma-C5; PCDHGA; pcdhga4; protocadherin 13; protocadherin 2; protocadherin 43; protocadherin gamma a1; protocadherin gamma A12; protocadherin gamma a2; protocadherin gamma B2; protocadherin gamma B6; protocadherin gamma b7; protocadherin gamma C-VI; protocadherin gamma c5; protocadherin gamma subfamily A, 10; protocadherin gamma subfamily A, 12; protocadherin gamma subfamily A, 3; protocadherin gamma subfamily A, 4; protocadherin gamma subfamily B, 1, isoform 2; protocadherin gamma subfamily B, 5; protocadherin gamma subfamily B, 7; protocadherin gamma subfamily C, 3; Protocadherin gamma-A1; Protocadherin gamma-A10; Protocadherin gamma-A11; Protocadherin gamma-A12; Protocadherin gamma-A2; Protocadherin gamma-A3; Protocadherin gamma-A4; Protocadherin gamma-A5; Protocadherin gamma-A6; protocadherin gamma-A6-like; Protocadherin gamma-A7; Protocadherin gamma-A8; Protocadherin gamma-A9; Protocadherin gamma-B1; Protocadherin gamma-B2; Protocadherin gamma-B3; Protocadherin gamma-B4; protocadherin gamma-B4-like; Protocadherin gamma-B5; Protocadherin gamma-B6; protocadherin gamma-B6-like; Protocadherin gamma-B7; Protocadherin gamma-C3; Protocadherin gamma-C4; Protocadherin gamma-C5; Protocadherin-2; Protocadherin-43

          View more View less

          Gene Aliases: CDH-GAMMA-A5; CDH20; CDH21; FIB2; FIB3; KIAA0327; KIAA0588; ME3; ME6; mKIAA0588; mKIAA4054; pc2c; PC43; PCDH-GAMMA-A1; PCDH-GAMMA-A10; PCDH-GAMMA-A11; PCDH-GAMMA-A12; PCDH-GAMMA-A2; PCDH-GAMMA-A3; PCDH-GAMMA-A4; PCDH-GAMMA-A5; PCDH-GAMMA-A6; PCDH-GAMMA-A7; PCDH-GAMMA-A8; PCDH-GAMMA-A9; PCDH-GAMMA-B1; PCDH-GAMMA-B2; PCDH-GAMMA-B3; PCDH-GAMMA-B4; PCDH-GAMMA-B5; PCDH-GAMMA-B6; PCDH-GAMMA-B7; PCDH-GAMMA-C3; PCDH-GAMMA-C4; PCDH-GAMMA-C5; Pcdh13; PCDH2; PCDHGA1; PCDHGA10; Pcdhga10temp; PCDHGA11; PCDHGA12; PCDHGA2; PCDHGA3; PCDHGA4; PCDHGA5; PCDHGA6; PCDHGA7; PCDHGA8; PCDHGA9; PCDHGB1; PCDHGB2; PCDHGB3; PCDHGB4; PCDHGB5; PCDHGB6; PCDHGB7; PCDHGC3; PCDHGC4; PCDHGC5; UNQ371/PRO707

          View more View less

          UniProt ID: (Human) Q9Y5H4, (Human) Q9Y5H3, (Human) Q9Y5H2, (Human) O60330, (Human) Q9Y5H1, (Human) Q9Y5H0, (Human) Q9Y5G9, (Human) Q9Y5G8, (Human) Q9Y5G7, (Human) Q9Y5G6, (Human) Q9Y5G5, (Human) Q9Y5G4, (Human) Q9Y5G3, (Human) Q9Y5G2, (Human) Q9Y5G1, (Human) Q9UN71, (Human) Q9Y5G0, (Human) Q9Y5F9, (Human) Q9Y5F8, (Human) Q9UN70, (Human) Q9Y5F7, (Human) Q9Y5F6, (Mouse) Q91XY4

          View more View less

          Entrez Gene ID: (Rat) 102552497, (Rat) 102552705, (Rat) 102552796, (Human) 56114, (Human) 56106, (Human) 56105, (Human) 26025, (Human) 56113, (Human) 56112, (Human) 56111, (Human) 56110, (Human) 56109, (Human) 56108, (Human) 9708, (Human) 56107, (Human) 56104, (Human) 56103, (Human) 56102, (Human) 8641, (Human) 56101, (Human) 56100, (Human) 56099, (Human) 5098, (Human) 56098, (Human) 56097, (Rat) 553129, (Mouse) 93709, (Rat) 498849, (Mouse) 93722, (Rat) 252897, (Mouse) 93723, (Mouse) 93724, (Rat) 498850, (Rat) 498846, (Mouse) 93710, (Mouse) 93711, (Rat) 498847, (Mouse) 93712, (Rat) 252894, (Rat) 291637, (Mouse) 93713, (Mouse) 93714, (Mouse) 93715, (Rat) 291635, (Rat) 364843, (Mouse) 93716, (Rat) 252895, (Mouse) 93717, (Mouse) 93699, (Mouse) 93700, (Mouse) 93701, (Mouse) 93702, (Rat) 680448, (Mouse) 93703, (Mouse) 93704, (Rat) 364844, (Mouse) 93706, (Rat) 116782, (Mouse) 93707, (Mouse) 93708, (Rat) 641525

          View more View less

          Function(s)
          calcium ion binding poly(A) RNA binding molecular_function protein binding
          Process(es)
          homophilic cell adhesion via plasma membrane adhesion molecules synapse organization cell adhesion spermatogenesis biological_process endosome to lysosome transport lipid tube assembly involved in organelle fusion calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0