Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Possible role in intracellular signaling and cytoskeleton dynamics at the Golgi.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PH domain-containing family G member 4; pleckstrin homology domain containing, family G (with RhoGef domain) member 4; Pleckstrin homology domain-containing family G member 4; Puratrophin-1; Purkinje cell atrophy associated protein 1; Purkinje cell atrophy-associated protein 1
Gene Aliases: 4931414L13Rik; AI448583; ARHGEF44; mFLJ00068; PLEKHG4; PRTPHN1; RGD1310790; SCA4
UniProt ID: (Human) Q58EX7
Entrez Gene ID: (Human) 25894, (Mouse) 102075, (Rat) 307796
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support