Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • QSOX1 Antibodies

          Proteintech

          QSOX1 Polyclonal Antibody

          19 References
          View all (13) QSOX1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite QSOX1 Polyclonal Antibody

          • Antibody Testing Data (12)
          QSOX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          QSOX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 12

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          QSOX1 Antibody (12713-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human heart tissue slide using 12713-1-AP ( QSOX1 Antibody) at dilution of 1:50 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          QSOX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QSOX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QSOX1 Antibody in Western Blot (WB)
          QSOX1 Antibody in Western Blot (WB)
          QSOX1 Antibody in Western Blot (WB)
          QSOX1 Antibody in Western Blot (WB)
          QSOX1 Antibody in Western Blot (WB)
          QSOX1 Antibody in Western Blot (WB)
          QSOX1 Antibody in Western Blot (WB)
          QSOX1 Antibody in Western Blot (WB)
          QSOX1 Antibody in Western Blot (WB)
          QSOX1 Antibody in Immunoprecipitation (IP)

          Product Details

          12713-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 9 publications 9 publications

          Immunohistochemistry (IHC)

          -
          View 8 publications 8 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Hamster, Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          QSOX1 Fusion Protein Ag3396 (1-329 aa encoded by BC017692)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.34 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MRRCNSGSG PPPSLLLLLL WLLAVPGANA APRSALYSPS DPLTLLQADT VRGAVLGSRS AWAVEFFASW CGHCIAFAPT WKALAEDVKA WRPALYLAAL DCAEETNSAV CRDFNIPGFP TVRFFKAFTK NGSGAVFPVA GADVQTLRER LIDALESHHD TWPPACPPLE PAKLEEIDGF FARNNEEYLA LIFEKGGSYL AREVALDLSQ HKGVAVRRVL NTEANVVRKF GVTDFPSCYL LFRNGSVSRV PVLMESRSFY TAYLQRLSGL TREAAQTTVA PTTANKIAPT VWKLADRSKI YMADLESALH YILRIEVGRF PVLEGQRLVA (1-329 aa encoded by BC017692)

          Target Information

          The QSOX1 gene, also known as Quiescin Q6, is a fusion of two ancient genes: thioredoxin and ERV1. Its expression is induced as fibroblasts begin to exit the proliferative cycle and enter quiescence, suggesting that this gene plays an important role in growth regulation. The QSOX1 protein oxidizes sulfhydryl groups to form disulfide bonds in proteins. QSOX1 expression is induced by hypoxia and appears to protect cells against oxidative stress-induced apoptosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: FAD-dependent sulfhydryl oxidase-2; hQSOX; mSOx; Quiescin Q6; quiescin Q6 sulfhydryl oxidase 1; RP11-502H18.3; rQSOX; Skin sulfhydryl oxidase; SOx-2; Sulfhydryl oxidase 1; testis tissue sperm-binding protein Li 62n; thiol oxidase 1

          View more View less

          Gene Aliases: 1300003H02Rik; Q6; QSCN6; QSOX; QSOX1; rSOx; SOx; Sox-2; Sox2; UNQ2520/PRO6013

          View more View less

          UniProt ID: (Human) O00391, (Mouse) Q8BND5, (Rat) Q6IUU3

          View more View less

          Entrez Gene ID: (Human) 5768, (Mouse) 104009, (Rat) 84491

          View more View less

          Function(s)
          protein disulfide isomerase activity flavin-linked sulfhydryl oxidase activity oxidoreductase activity thiol oxidase activity
          Process(es)
          protein folding negative regulation of macroautophagy cell redox homeostasis oxidation-reduction process platelet degranulation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0