Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTGTPGAVAT RDGEAPERSP PCSPSYDLTG KVMLLGDTGV GKTCFLIQFK DGAFLSGTFI ATVGIDFRNK VVTVDGVRVK LQIWDTAGQE RFRSVTHAYY RDAQALLLLY DITNKSSFDN IRAWLTEIHE YAQRDVVIML LGNKADMSSE RVIRSEDGET LAREYGVPFL ETSAKTGMNV ELAFLAIAKE LKYRAGHQAD EPSFQIRDYV ESQKKRSSCC SFM
Rab proteins are low molecular mass GTPases that are critical regulators of vesicle trafficking. For additional background information on Rab proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RAB37, member of RAS oncogene family; Ras-related protein Rab-37
Gene Aliases: RAB37
UniProt ID: (Human) Q96AX2
Entrez Gene ID: (Human) 326624
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support