Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AQPVQDTKL HVFDANTLTW SQPETLGNPP SPRHGHVMVA AGTKLFIHGG LAGDRFYDDL HCIDISDMKW QKLNPTGAAP AGCAAHSAVA MGKHVYIFGG MTPAGALDTM YQYHTEEQHW TLLKFDTLLP PGRLDHSMCI IPWPVTCASE KEGSNSLTLN HEAEKEDSAD KVMSHSGDSH EESQTATLLC LVFGG (162-355 aa encoded by BC000503)
The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells. This cytokine stimulates cell proliferation and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. The gene encoding this cytokine has been identified as a candidate gene for asthma. Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40 kDa Rab9 effector protein; p40; Rab9 effector p40; Rab9 effector protein with kelch motifs
Gene Aliases: 8430412M01Rik; 9530020D24Rik; AV073337; bA65N13.1; C87311; p40; RAB9P40; RABEPK; RGD1310612
UniProt ID: (Human) Q7Z6M1, (Rat) Q4V8F4, (Mouse) Q8VCH5
Entrez Gene ID: (Human) 10244, (Rat) 296649, (Mouse) 227746
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support