Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • RACK1 Antibodies

          Invitrogen

          RACK1 Polyclonal Antibody

          Advanced Verification
          View all (19) RACK1 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite RACK1 Polyclonal Antibody

          • Antibody Testing Data (6)
          • Advanced Verification (2)
          RACK1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RACK1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RACK1 Antibody (PA5-79319) in ICC/IF

          Immunocytochemistry analysis of RACK1 using anti-RACK1 antibody (Product # PA5-79319). RACK1 was detected in a section of A431 cells. Enzyme antigen retrieval was performed using IHC enzyme antigen retrieval reagent for 15 mins. The cells were blocked with 10% goat serum and then incubated with 2μg/mL rabbit anti-RACK1 antibody (Product # PA5-79319)overnight at 4°C. DyLight®550 Conjugated Goat Anti-Rabbit IgG was used as secondary antibody at 1:100 dilution and incubated for 30 minutes at 37°C. The section was counterstained with DAPI. Vis... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RACK1 Antibody in Immunocytochemistry (ICC/IF)
          RACK1 Antibody in Immunocytochemistry (ICC/IF)
          RACK1 Antibody in Western Blot (WB)
          RACK1 Antibody in Western Blot (WB)
          RACK1 Antibody in Western Blot (WB)
          RACK1 Antibody in Flow Cytometry (Flow)
          RACK1 Antibody
          RACK1 Antibody

          Product Details

          PA5-79319

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.5 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          2 µg/mL
          -

          Flow Cytometry (Flow)

          1-3 µg/1x10^6 cells
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived human RACK1 recombinant protein (Position: T2-R317).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          500 µg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 4mg trehalose

          Contains

          0.05mg sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2746435

          Product Specific Information

          Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

          Positive Control - WB: human MCF-7 whole cell, human Jurkat whole cell, human HepG2 whole cell, human A549 whole cell, human Raji whole cell, rat liver tissue, mouse RAW2647 whole cell, mouse HEPA1-6 whole cell. ICC/IF: A431 cell. Flow: A431 cell.

          Target Information

          RACK1 (receptor for activated C kinase 1) was identified through its binding to various PKC isoforms. Its main function is to recruit PKC and various other proteins to specific locations to form multiprotein complexes, mediating various signal pathways. RACK1 is required for VANGL2 membrane localization and for PTK2/FAK1 phosphorylation and dephosphorylation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 12-3; activated protein kinase C receptor; Cell proliferation-inducing gene 21 protein; guanine nucleotide binding protein (G protein) beta polypeptide 2-like 1; guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1 sequence 1; guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1; guanine nucleotide binding protein beta polypeptide 2-like 1; guanine nucleotide binding protein related; guanine nucleotide binding protein, beta 2, related sequence 1; guanine nucleotide binding protein, beta polypeptide 2-like 1; guanine nucleotide binding protein, beta-2, related sequence 1; Guanine nucleotide-binding protein subunit beta-2-like 1; Guanine nucleotide-binding protein subunit beta-like protein 12.3; HLC-7; Human lung cancer oncogene 7 protein; lung cancer oncogene 7; OTTHUMP00000223870; OTTHUMP00000223891; OTTHUMP00000223893; OTTHUMP00000223900; OTTHUMP00000223901; OTTHUMP00000223902; OTTHUMP00000223930; OTTHUMP00000223931; p205; proliferation-inducing gene 21; protein homologous to chicken B complex protein, guanine nucleotide binding; protein kinase C receptor; rack-1; Receptor for activated C kinase; receptor for activated protein kinase C; Receptor of activated protein C kinase 1; Receptor of activated protein kinase C 1; Small ribosomal subunit protein RACK1

          View more View less

          Gene Aliases: AL033335; GB-like; Gnb2-rs1; GNB2L1; H12.3; HLC-7; HLC7; p205; PIG21; RACK1

          View more View less

          UniProt ID: (Rat) P63245, (Mouse) P68040, (Human) P63244

          View more View less

          Entrez Gene ID: (Rat) 83427, (Mouse) 14694, (Human) 10399

          View more View less

          Function(s)
          protein kinase C binding ion channel inhibitor activity cysteine-type endopeptidase activator activity involved in apoptotic process enzyme binding protein phosphatase binding protein tyrosine kinase inhibitor activity receptor tyrosine kinase binding SH2 domain binding protein homodimerization activity poly(A) RNA binding receptor activity protein binding signaling adaptor activity receptor binding protein complex scaffold cadherin binding involved in cell-cell adhesion
          Process(es)
          osteoblast differentiation positive regulation of protein phosphorylation activation of cysteine-type endopeptidase activity involved in apoptotic process protein kinase C-activating G-protein coupled receptor signaling pathway gastrulation negative regulation of gene expression negative regulation of translation negative regulation of Wnt signaling pathway negative regulation of cell growth positive regulation of cell migration positive regulation of cAMP catabolic process positive regulation of proteasomal ubiquitin-dependent protein catabolic process positive regulation of protein homooligomerization regulation of protein localization positive regulation of apoptotic process positive regulation of GTPase activity rhythmic process negative regulation of phagocytosis regulation of cell division positive regulation of cyclic-nucleotide phosphodiesterase activity regulation of cell cycle negative regulation of protein kinase B signaling positive regulation of mitochondrial depolarization negative regulation of protein tyrosine kinase activity regulation of establishment of protein localization to plasma membrane negative regulation of hydrogen peroxide-induced neuron death regulation of establishment of cell polarity positive regulation of gastrulation positive regulation of intrinsic apoptotic signaling pathway regulation of translation apoptotic process cell cycle multicellular organism development protein localization negative regulation of peptidyl-serine phosphorylation intracellular signal transduction regulation of growth cellular response to glucose stimulus regulation of tumor necrosis factor-mediated signaling pathway viral process positive regulation of Golgi to plasma membrane protein transport cellular response to growth factor stimulus cell-cell adhesion negative regulation of endoplasmic reticulum unfolded protein response positive regulation of ceramide biosynthetic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67c77f6dc7-n6nf2:80/100.66.135.186:80.
          git-commit: 545e792480003146e539d624bd2a21b51d198228
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0