Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
RAPGEF6 is a guanine nucleotide exchange factor (GEF) for Rap1A, Rap2A and M-Ras GTPases. RAPGEF6 does not interact with cAMP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PDZ domain containing guanine nucleotide exchange factor (GEF) 2; PDZ domain containing guanine nucleotide exchange factor(GEF)1; PDZ domain-containing guanine nucleotide exchange factor 2; PDZ domain-containing guanine nucleotide exchange factor I; PDZ-GEF2; RA-GEF-2; Rap guanine nucleotide exchange factor (GEF) 6; rap guanine nucleotide exchange factor 2; Rap guanine nucleotide exchange factor 6
Gene Aliases: A530068K01; AI844632; BB085664; C030018K18Rik; KIA001LB; mKIAA4052; PDZ-GEF2; PDZGEF2; RA-GEF-2; RAGEF2; RAPGEF6
UniProt ID: (Human) Q8TEU7
Entrez Gene ID: (Human) 51735, (Rat) 303141, (Mouse) 192786
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support