Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
RALA, resorcylic acid lactone alpha, is a low molecular weight GTP binding protein belonging to the RAS family of onco proteins. RALA has a 50% amino acid homology with RAS and an 85% homology with RALB. GTP binding proteins mediate the transmembrane signaling initiated by the occupancy of certain cell surface receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: -ral simian leukemia viral oncogene homolog A (ras related); GTP binding protein); ral-A protein; Ras family small GTP binding protein RALA; ras related GTP binding protein A; ras related GTP binding protein B; RAS-like protein A; RAS-like protein B; RAS-like, family 1; Ras-related protein Ral-A; Ras-related protein Ral-B; v-ral simian leukemia viral oncogene homolog A (ras related); v-ral simian leukemia viral oncogene homolog B (ras related); v-ral simian leukemia viral oncogene homolog B (ras related; GTP binding protein)
Gene Aliases: 3010001O15Rik; 5730472O18Rik; AW322615; dRalb; RAL; Ral-a; Ral-b; RALA; RALB; Rasl1
UniProt ID: (Human) P11233, (Human) P11234, (Mouse) P63321, (Rat) P63322, (Rat) P36860, (Mouse) Q9JIW9
Entrez Gene ID: (Human) 5898, (Human) 5899, (Mouse) 56044, (Rat) 81757, (Rat) 116546, (Mouse) 64143
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support