Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • IPO5 Antibodies

          Abnova

          RANBP5 Polyclonal Antibody, MaxPab™

          View all (10) IPO5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite RANBP5 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          RANBP5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RANBP5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RANBP5 Antibody (H00003843-B01P) in WB

          RANBP5 MaxPab polyclonal antibody. Western Blot analysis of RANBP5 expression in HeLa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RANBP5 Antibody in Western Blot (WB)

          Product Details

          H00003843-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RANBP5 (AAH45640.1, 1 a.a. approximately 1115 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MPEDQVGKLE ATENTISAMA AAAAEQQQFY LLLGNLLSPD NVVRKQAEET YENIPGQSKI TFLLQAIRNT TAAEEARQMA AVLLRRLLSS AFDEVYPALP SDVQTAIKSE LLMIIQMETQ SSMRKKVCDI AAELARNLID EDGNNQWPEG LKFLFDSVSS QNVGLREAAL HIFWNFPGIF GNQQQHYLDV IKRMLVQCMQ DQEHPSIRTL SARATAAFIL ANEHNVALFK HFADLLPGFL QAVNDSCYQN DDSVLKSLVE IADTVPKYLR PHLEATLQLS LKLCGDTSLN NMQRQLALEV IVTLSETAAA MLRKHTNIVA QTIPQMLAMM VDLEEDEDWA NADELEDDDF DSNAVAGESA LDRMACGLGG KLVLPMIKEH IMQMLQNPDW KYRHAGLMAL SAIGEGCHQQ MEGILNEIVN FVLLFLQDPH PRVRYAACNA VGQMATDFAP GFQKKFHEKV IAALLQTMED QGNQRVQAHA AAALINFTED CPKSLLIPYL DNLVKHLHSI MVLKLQELIQ KGTKLVLEQV VTSIASVADT AEEKFVPYYD LFMPSRKHIV ENAVQKELRL LRGKTIECIS LIGLAVGKEK FMQDASDVMQ LLLKTQTDFN DMEDDDPQIS YMISAWARMC KILGKEFQQY LPVVMGPLMK TASIKPEVAL LDTQDMENMS DDDGWEFVNL GDQQSFGIKT AGLEEKSTAC QMLVCYAKEL KEGFVEYTEQ VVKLMVPLLK FYFHDGVRVA AAESMPLLLE CARVRGPEYL TQMWHFMCDA LIKAIGTEPD SDVLSEIMHS FAKCIEVMGD GCLNNEHFEE LGGILKAKLE EHFKNQELRQ VKRQDEDYDE QVEESLQDED DNDVYILTKV SDILHSIFSS YKEKVLPWFE QLLPLIVNLI CPHRPWPDRQ WGLCIFDDVI EHCSPASFKY AEYFLRPMLQ YVCDNSPEVR QAAAYGLGVM AQYGGDNYRP FCTEALPLLV RVIQSADSKT KENVNATENC ISAVGKIMKF KPDCVNVEEV LPHWLSWLPL HEDKEEAVQT FNYLCDLIES NHPIVLGPNN TNLPKIFSII AEGEMHEAIK HEDPCAKRLA NVVRQVQTSG GLWTECIAQL SPEQQAAIQE LLNSA

          Target Information

          Nucleocytoplasmic transport, a signal- and energy-dependent process, takes place through nuclear pore complexes embedded in the nuclear envelope. The import of proteins containing a nuclear localization signal (NLS) requires the NLS import receptor, a heterodimer of importin alpha and beta subunits also known as karyopherins. Importin alpha binds the NLS-containing cargo in the cytoplasm and importin beta docks the complex at the cytoplasmic side of the nuclear pore complex. In the presence of nucleoside triphosphates and the small GTP binding protein Ran, the complex moves into the nuclear pore complex and the importin subunits dissociate. Importin alpha enters the nucleoplasm with its passenger protein and importin beta remains at the pore. Interactions between importin beta and the FG repeats of nucleoporins are essential in translocation through the pore complex. The protein encoded by this gene is a member of the importin beta family.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Imp5; importin 5; importin beta-3 subunit; Importin subunit beta-3; Importin-5; IPO 5; karyopherin (importin) beta 3; Karyopherin beta 3; Karyopherin beta-3; MGC2068; RAN binding protein 5; Ran-binding protein 5; Ran_GTP binding protein 5; RanBP 5; RanBP5

          View more View less

          Gene Aliases: IMB3; imp5; IPO5; KPNB3; Pse1; RANBP5

          View more View less

          UniProt ID: (Human) O00410

          View more View less

          Entrez Gene ID: (Human) 3843, (Rat) 306182

          View more View less

          Function(s)
          GTPase inhibitor activity protein binding nuclear localization sequence binding Ran GTPase binding protein transporter activity poly(A) RNA binding
          Process(es)
          NLS-bearing protein import into nucleus ribosomal protein import into nucleus intracellular protein transport positive regulation of protein import into nucleus cellular response to amino acid stimulus protein import into nucleus, docking protein import into nucleus, translocation viral process negative regulation of GTPase activity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0