Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASEDIAKLA ETLAKTQVAG GQLSFKGKSL KLNTAEDAKD VIKEIEDFDS LEALRLEGNT VGVEAARVIA KALEKKSELK RCHWSDMFTG RLRTEIPPAL ISLGEGLITA GAQLVELDLS DNAFGPDGVQ GFEALLKSSA CFTLQELKLN NCGMGIGGGK ILAAALTECH RKSSAQGKPL ALKVFVAGRN RLENDGATAL AEAFRVIGTL EEVHMPQNGI NHPGITALAQ AFAVNPLLRV INLNDNTFTE KGAVAMAETL KTLRQVEVIN FGDCLVRSKG AVAIADAIRG GLPKLKELNL SFCEIKRDAA LAVAEAMADK AELEKLDLNG NTLGEEGCEQ LQEVLEGFNM AKVLASLSDD EDEEEEEEGE EEEEEAEEEE EEDEEEEEEE EEEEEEEPQQ RGQGEKSATP SRKILDPNTG EPAPVLSSPP PADVSTFLAF PSPEKLLRLG PKSSVLIAQQ TDTSDPEKVV SAFLKVSSVF KDEATVRMAV QDAVDALMQK AFNSSSFNSN TFLTRLLVHM GLLKSEDKVK AIANLYGPLM ALNHMVQQDY FPKALAPLLL AFVTKPNSAL ESCSFARHSL LQTLYKV
Ran is a small signaling GTPase that is involved in nucleocytoplasmic transport. Two additional functions of animal Ran in the formation of spindle asters and the reassembly of the nuclear envelope in mitotic cells have been recently reported. In contrast to Ras or Rho, Ran is not associated with membranes. Instead, the spatial sequestering of its accessory proteins, the Ran GTPase-activating protein RanGAP and the nucleotide exchange factor RCC1, appears to define the local concentration of RanGTP vs. RanGDP involved in signaling. Mammalian RanGAP is bound to the nuclear pore by a mechanism involving the attachment of small ubiquitin-related modifier protein (SUMO) to its C terminus and the subsequent binding of the SUMOylated domain to the nucleoporin Nup358.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC20266; Ran GTPase-activating protein 1; RanGAP1; segregation distorter homolog; segregation distortion
Gene Aliases: Fug1; KIAA1835; RANGAP; RANGAP1; SD
UniProt ID: (Human) P46060
Entrez Gene ID: (Human) 5905
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support