Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: QLINMGVIP TLVKLLGIHC QNAALTEMCL VAFGNLAELE SSKEQFASTN IAEELVKLFK KQIEHDKREM IFEVLAPLAE NDAIKLQLVE AGLVECLLEI VQQKVDSDKE DDITELKTGS DLMVLLLLGD ESMQKLFEGG KGSVFQRVLS WIPSNNHQLQ LAGALAIANF ARNDANCIHM VDNGIVEKLM DLLDRHVEDG NVTVQHAALS ALRNLAIPVI NKAKMLSAGV TE (176-406 aa encoded by BC001851)
The smg GDP dissociation stimulator (smgGDS) protein is a stimulatory GDP/GTP exchange protein with GTPase activity (Riess et al., 1993 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exchange factor smgGDS; Rap1 GTPase-GDP dissociation stimulator 1; RAP1, GTP-GDP dissociation stimulator 1; SMG GDS protein; SMG P21 stimulatory GDP/GTP exchange protein
Gene Aliases: BC011279; GDS1; RAP1GDS1; SMGGDS
UniProt ID: (Human) P52306, (Mouse) E9Q912
Entrez Gene ID: (Human) 5910, (Mouse) 229877
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support