Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MREYKVVVL GSGGVGKSAL TVQFVTGSFI EKYDPTIEDF YRKEIEVDSS PSVLEILDTA GTEQFASMRD LYIKNGQGFI LVYSLVNQQS FQDIKPMRDQ IIRVKRYERV PMILVGNKVD LEGEREVSYG EGKALAEEWS CPFMETSAKN KASVDELFAE IVRQMNYAAQ PNGDEGCCSA CVIL (1-183 aa encoded by BC012362)
This intronless gene belongs to a family of RAS-related genes. The proteins encoded by these genes share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. The most striking difference between the RAP and RAS proteins resides in their 61st amino acid: glutamine in RAS is replaced by threonine in RAP proteins. Evidence suggests that this protein may be polyisoprenylated and palmitoylated.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ras-related protein Rap-2b; small GTP binding protein
Gene Aliases: 4021402C18Rik; AA408554; RAP2B
UniProt ID: (Human) P61225, (Rat) P61227, (Mouse) P61226
Entrez Gene ID: (Human) 5912, (Rat) 170923, (Mouse) 74012
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support