Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • RARA Antibodies

          Invitrogen

          RARA Recombinant Rabbit Monoclonal Antibody (SN0725)

          1 Reference
          View all (38) RARA antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite RARA Recombinant Rabbit Monoclonal Antibody (SN0725)

          • Antibody Testing Data (1)
          RARA Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          RARA Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RARA Antibody (MA5-32325) in Flow

          Flow Cytometric analysis of RARA in MCF-7 cells using a RARA Monoclonal Antibody (Product # MA5-32325) at a dilution of 1:50, as seen in red compared with an unlabelled control (cells without incubation with primary antibody; black). Alexa Fluor 488-conjugated goat anti rabbit IgG was used as the secondary antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RARA Antibody in Flow Cytometry (Flow)

          Product Details

          MA5-32325

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:50-1:2,000
          -

          Flow Cytometry (Flow)

          1:50-1:100
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          SN0725

          Immunogen

          Synthetic peptide within Human Retinoic Acid Receptor alpha aa 1-50
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 40% Glycerol, 0.05% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2809606

          Product Specific Information

          Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

          Target Information

          RARA is a receptor for retinoic acid. Retinoic acid receptors bind as heterodimers to their target response elements in response to their ligands, all-trans or 9-cis retinoic acid, and regulate gene expression in various biological processes. The RXR/RAR heterodimers bind to the retinoic acid response elements (RARE) composed of tandem 5'-AGGTCA-3' sites known as DR1-DR5. In the absence of ligand, the RXR-RAR heterodimers associate with a multiprotein complex containing transcription corepressors that induce histone acetylation, chromatin condensation and transcriptional suppression. On ligand binding, the corepressors dissociate from the receptors and associate with the coactivators leading to transcriptional activation. RARA plays an essential role in the regulation of retinoic acid-induced germ cell development during spermatogenesis. Has a role in the survival of early spermatocytes at the beginning prophase of meiosis. In Sertoli cells, may promote the survival and development of early meiotic prophase spermatocytes. In concert with RARG, required for skeletal growth, matrix homeostasis and growth plate function. Regulates expression of target genes in a ligand-dependent manner by recruiting chromatin complexes containing KMT2E/MLL5. Mediates retinoic acid-induced granulopoiesis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Nuclear receptor subfamily 1 group B member 1; nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form; RAR Alpha; RAR alpha 1; RAR-alpha; retinoic acid; retinoic acid nuclear receptor alpha variant 1; retinoic acid nuclear receptor alpha variant 2; retinoic acid receptor; Retinoic acid receptor alpha; retinoic acid receptor, alpha polypeptide; RRA

          View more View less

          Gene Aliases: NR1B1; RAR; RARA; RARalpha1

          View more View less

          UniProt ID: (Human) P10276, (Mouse) P11416

          View more View less

          Entrez Gene ID: (Human) 5914, (Mouse) 19401

          View more View less

          Function(s)
          translation repressor activity, nucleic acid binding RNA polymerase II regulatory region sequence-specific DNA binding retinoic acid binding transcription factor activity, sequence-specific DNA binding steroid hormone receptor activity retinoic acid receptor activity transcription coactivator activity transcription corepressor activity receptor binding protein binding transcription factor binding drug binding zinc ion binding enzyme binding protein domain specific binding chromatin DNA binding histone deacetylase binding protein kinase B binding retinoic acid-responsive element binding protein heterodimerization activity mRNA 5'-UTR binding protein kinase A binding alpha-actinin binding regulatory region DNA binding RNA polymerase II regulatory region DNA binding DNA binding RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding phosphatidylinositol 3-kinase regulator activity sequence-specific DNA binding transcription regulatory region DNA binding metal ion binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter ureteric bud development neural tube closure glandular epithelial cell development outflow tract septum morphogenesis growth plate cartilage development transcription, DNA-templated regulation of transcription, DNA-templated transcription from RNA polymerase II promoter protein phosphorylation signal transduction germ cell development spermatogenesis positive regulation of cell proliferation negative regulation of cell proliferation male gonad development positive regulation of gene expression negative regulation of gene expression positive regulation of phosphatidylinositol 3-kinase signaling negative regulation of translation regulation of granulocyte differentiation negative regulation of granulocyte differentiation embryonic camera-type eye development regulation of myelination response to retinoic acid negative regulation of interferon-gamma production negative regulation of tumor necrosis factor production positive regulation of interleukin-13 production positive regulation of interleukin-4 production positive regulation of interleukin-5 production multicellular organism growth regulation of apoptotic process negative regulation of apoptotic process apoptotic cell clearance steroid hormone mediated signaling pathway regulation of phosphatidylinositol 3-kinase activity response to ethanol negative regulation of cell differentiation positive regulation of T-helper 2 cell differentiation positive regulation of neuron differentiation positive regulation of cell cycle negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of translational initiation regulation of synaptic plasticity retinoic acid receptor signaling pathway positive regulation of binding positive regulation of protein kinase B signaling ventricular cardiac muscle cell differentiation Sertoli cell fate commitment limb development face development bone development trachea cartilage development chondroblast differentiation negative regulation of cartilage development positive regulation of ERK1 and ERK2 cascade cellular response to lipopolysaccharide cellular response to retinoic acid cellular response to estrogen stimulus liver development transcription initiation from RNA polymerase II promoter female pregnancy hippocampus development prostate gland development response to estradiol response to vitamin A response to cytokine
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0