Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DGLRSALEN SYSKFNRSLH INNISPGNTI SRSSPKFIGH SPQPLGYDRS SRSSYVDEDC DSLALCDPLQ RTNCEVDSLP ESCFDSGLST LRDPNEYDSE VEYKHQRGFQ RSHGVQESFG GDASDTDVPD IRDEETFGLE DVASVLDWKP QGSVSEGSIV SSSRKPISAL SPQTDLVDDN AKSFSSQKAY KIVLAGDAAV GKSSFLMRLC KNEFRENISA TLGVDFQMKT LIVDGERTVL QLWDTAGQER FRSIAKSYFR KADGVLLLYD VTCEKSFLNI REWVDMIEDA AHETVPIMLV GNKADIRDTA ATEGQKCVPG HFGEKLAMTY GALFCETSAK DGSNIVEAVL HLAREVKKRT DKDDSRSITN LTGTNSKKSP QMKNCCNG (354-740 aa encoded by BC023566)
Binds predominantly GDP, and also GTP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ras and EF-hand domain-containing protein; Ras and EF-hand domain-containing protein homolog; Ras-related protein Rab-45
Gene Aliases: BC030882; RAB45; RASEF
UniProt ID: (Human) Q8IZ41, (Mouse) Q5RI75
Entrez Gene ID: (Human) 158158, (Mouse) 242505
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support