Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEADGDGEE LARLRSVFAA CDANRSGRLE REEFRALCTE LRVRPADAEA VFQRLDADRD GAITFQEFAR GFLGSLRGGR RRDWGPLDPA PAVSEAGPET HDSEEDEGDE DAAAALATSC GPASPGRAWQ DFQARLGDEA KFIPREEQVS TLYQNINLVE PRLIQPYEHV IKNFIREIRL QSTEMENLAI AVKRAQDKAA MQLSELEEEM DQRIQAAEHK TRKDEKRKAE EALSDLRRQY ETEVGD (1-245 aa encoded by BC023566)
Binds predominantly GDP, and also GTP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ras and EF-hand domain-containing protein; Ras-related protein Rab-45
Gene Aliases: RAB45; RASEF
UniProt ID: (Human) Q8IZ41
Entrez Gene ID: (Human) 158158, (Rat) 298138
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support