Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: GTLDLDKGCT VEELLRGCIE AFDDSGKVRD PQLVRMFLMM HPWYIPSSQL AAKLLHIYQQ SRKDNSNSLQ VKTCHLVRYW ISAFPAEFDL NPELAEQIKE
The protein encoded by this gene is a brain-enriched nucleotide exchanged factor that contains an N-terminal GEF domain, 2 tandem repeats of EF-hand calcium-binding motifs, and a C-terminal diacylglycerol/phorbol ester-binding domain. This protein can activate small GTPases, including RAS and RAP1/RAS3. The nucleotide exchange activity of this protein can be stimulated by calcium and diacylglycerol. Three alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium and DAG-regulated guanine nucleotide exchange factor I; calcium and diacylglycerol-regulated guanine nucleotide exchange factor I; CalDAG-GEFI; Cdc25-like protein; F25B3.3 kinase-like protein; guanine exchange factor MCG7; hCDC25L; RAS guanyl nucleotide-releasing protein 2; RAS guanyl releasing protein 2 (calcium and DAG-regulated); RAS guanyl-releasing protein 2
Gene Aliases: CALDAG-GEFI; CDC25L; MCG7; RASGRP2
UniProt ID: (Human) Q7LDG7
Entrez Gene ID: (Human) 10235
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support