Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • RASSF1 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Invitrogen

          RASSF1A Monoclonal Antibody (eB114-10H1), eBioscience™

          Advanced Verification
          22 Published Figures
          21 References
          View all (28) RASSF1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Datasheet
          Protocols

          Cite RASSF1A Monoclonal Antibody (eB114-10H1), eBioscience™

          • Published Figures (22)
          • Advanced Verification (1)
          RASSF1A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RASSF1A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 23

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RASSF1A Antibody (14-6888-37) in ICC/IF

          The Biochemical journal 2010 - Figure 3 Analysis of mitotic signalling proteins in RASSF7 -knockdown HeLa cells ( A ) Active PLK1 (green) appeared normal. ( B ) RASSF1A (green) appeared normal. ( C and D ) Aurora A (red, arrows) and Aurora B (red, arrowheads) appeared to localize normally. Phospho-Aurora at the centrosomes (green, arrows) appeared normal, but phospho-Aurora at the kinetochore (green, arrowheads) was reduced (9.3% of cell... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RASSF1A Antibody in Immunocytochemistry (ICC/IF)
          RASSF1A Antibody in Immunohistochemistry (IHC)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot, Immunoprecipitation (WB, IP)
          RASSF1A Antibody in Western Blot, Immunoprecipitation (WB, IP)
          RASSF1A Antibody in Western Blot, Immunoprecipitation (WB, IP)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot, Immunoprecipitation (WB, IP)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody in Western Blot (WB)
          RASSF1A Antibody

          Product Details

          14-6888-37

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-10 µg/mL
          View 14 publications 14 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          Assay-Dependent
          View 3 publications 3 publications
          Product Specifications

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eB114-10H1

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Storage conditions

          4°C

          Shipping conditions

          Wet ice

          RRID

          AB_2865247

          Product Specific Information

          Description: The eB114 antibody reacts with the RASSF1A isoform of the tumor suppressor gene RASSF1. RASSF1A contains a Ras Association Domain (RA) and binds Ras in a GTP-dependent manner to effect apoptosis. The RASSF1A isoform undergoes epigenetic inactivation in a variety of human primary tumors. This has been associated with hypermethylation of a CpG island(s) in its promoter region. This antibody was generated against a synthetic peptide corresponding to the splice junction region of RASSF1A.

          Applications Reported: The eB114 antibody has been reported for use in immunoprecipitation, and immunoblotting (WB).

          Applications Tested: The eB114-10H1 has been tested by Western blotting and can be used at approximately 1-10 µg/mL. RASS1A is detected as a 45 kDa band in Western blotting of a HeLa cell extract. It is recommended that the reagent be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          This gene encodes a protein similar to the RAS effector proteins. Loss or altered expression of this gene has been associated with the pathogenesis of a variety of cancers, which suggests the tumor suppressor function of this gene. The inactivation of this gene was found to be correlated with the hypermethylation of its CpG-island promoter region. The encoded protein was found to interact with DNA repair protein XPA. The protein was also shown to inhibit the accumulation of cyclin D1, and thus induce cell cycle arrest. Seven alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0