Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • FIP200 Antibodies

          Proteintech

          RB1CC1 Polyclonal Antibody

          98 References
          View all (14) FIP200 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite RB1CC1 Polyclonal Antibody

          • Antibody Testing Data (17)
          RB1CC1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RB1CC1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 17

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RB1CC1 Antibody (17250-1-AP) in ICC/IF

          Immunofluorescent analysis of MCF-7 cells, using RB1CC1 antibody 17250-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IgG (red). Blue pseudocolor = DAPI (fluorescent DNA dye). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RB1CC1 Antibody in Immunocytochemistry (ICC/IF)
          RB1CC1 Antibody in Immunocytochemistry (ICC/IF)
          RB1CC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RB1CC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RB1CC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RB1CC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RB1CC1 Antibody in Western Blot (WB)
          RB1CC1 Antibody in Western Blot (WB)
          RB1CC1 Antibody in Western Blot (WB)
          RB1CC1 Antibody in Western Blot (WB)
          RB1CC1 Antibody in Western Blot (WB)
          RB1CC1 Antibody in Western Blot (WB)
          RB1CC1 Antibody in Western Blot (WB)
          RB1CC1 Antibody in Western Blot (WB)
          RB1CC1 Antibody in Western Blot (WB)
          RB1CC1 Antibody in Immunoprecipitation (IP)
          RB1CC1 Antibody in Immunoprecipitation (IP)

          Product Details

          17250-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:6,000
          View 58 publications 58 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 28 publications 28 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 4 publications 4 publications

          Miscellaneous PubMed (MISC)

          -
          View 2 publications 2 publications

          Not applicable (N/A)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RB1CC1 Fusion Protein Ag10508 (1241-1594 aa encoded by BC017556)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human RB1CC1. the calculated molecular weight of RB1CC1 is 180 kDa, but the modified RB1CC1 is about 200 kDa.

          Immunogen sequence: AIQTALKEF KLEREVVEKE LLEKVKHLEN QIAKSPAIDS TRGDSSSLVA ELQEKLQEEK AKFLEQLEEQ EKRKNEEMQN VRTSLIAEQQ TNFNTVLTRE KMRKENIIND LSDKLKSTMQ QQERDKDLIE SLSEDRARLL EEKKKLEEEV SKLRSSSFVP SPYVATAPEL YGACAPELPG ESDRSAVETA DEGRVDSAME TSMMSVQENI HMLSEEKQRI MLLERTLQLK EEENKRLNQR LMSQSMSSVS SRHSEKIAIR DFQVGDLVLI ILDERHDNYV LFTVSPTLYF LHSESLPALD LKPGEGASGA SRRPWVLGKV MEKEYCQAKK AQNRFKVPLG TKFYRVKAVS WNKKV (1241-1594 aa encoded by BC017556 )

          Target Information

          The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 200 kDa FAK family kinase-interacting protein; coiled coil forming protein 1; Coiled-coil-forming protein 1; FAK family kinase-interacting protein of 200 kDa; FIP200; LaXp180; phosphatase 1, regulatory subunit 131; RB1-inducible coiled-coil 1; RB1-inducible coiled-coil protein 1

          View more View less

          Gene Aliases: 2900055E04Rik; 5930404L04Rik; ATG17; CC1; FIP200; KIAA0203; LaXp180; PPP1R131; RB1CC1; RBICC

          View more View less

          UniProt ID: (Human) Q8TDY2, (Mouse) Q9ESK9

          View more View less

          Entrez Gene ID: (Human) 9821, (Mouse) 12421, (Rat) 312927

          View more View less

          Function(s)
          protein binding protein kinase binding molecular_function
          Process(es)
          autophagosome assembly liver development positive regulation of protein phosphorylation transcription, DNA-templated regulation of transcription, DNA-templated autophagy cell cycle JNK cascade heart development regulation of autophagy negative regulation of apoptotic process positive regulation of cell size negative regulation of extrinsic apoptotic signaling pathway biological_process macroautophagy
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0