Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSCVHYKFSS KLNYDTVTFD GLHISLCDLK KQIMGREKLK AADCDLQITN AQTKEEYTDD NALIPKNSSV IVRRIPIGGV KSTSKTYVIS RTEPAMATTK AVCKNTISHF FYTLLLPL
This gene encodes a cilia-associated protein belonging to the kinesin family. This protein plays a role in the sonic hedgehog (SHH) signaling pathway through the regulation of GLI transcription factors. It functions as a negative regulator of the SHH pathway by preventing inappropriate activation of GLI2 in the absence of ligand, and as a positive regulator by preventing the processing of GLI3 into its repressor form. Mutations in this gene have been associated with various ciliopathies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase RBBP6; p53-associated cellular protein of testis; Proliferation potential-related protein; Protein P2P-R; RB-binding Q-protein 1; RBQ-1; retinoblastoma binding protein 6; Retinoblastoma-binding protein 6; retinoblastoma-binding protein 6, isoform 3; Retinoblastoma-binding Q protein 1; RING-type E3 ubiquitin transferase RBBP6
Gene Aliases: MY038; P2P-R; P2PR; PACT; RBBP6; RBQ-1; RBQ1; SNAMA
UniProt ID: (Human) Q7Z6E9
Entrez Gene ID: (Human) 5930
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support