Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSHRAKLAMS GKIIIRNPIK IGYGKATPTT RLWVGGLGPW VPLAALAREF DRFGTIRTID YRKGDSWAYI QYESLDAAHA AWTHMRGFPL GGPDRRLRVD FADTEHRYQQ QYLQPLPLTH YELVTDAFGH RAPDPLRGAR DRTPPLLYRD RDRDLYPDSD WVPPPPPVRE RSTRTAATSV PAYEPLDSLD RRRDGWSLDR DRGDRDLPSS RDQPRKRRLP EESGGRHLDR SPESDRPRKR HCAPSPDRSP ELSSSRDRYN SDNDRSSRLL LERPSPIRDR RGSLEKSQGD KRDRKNSASA ERDRKHRTTA PTEGKSPLKK EDRSDGSAPS TSTASSKLKS PSQKQDGGTA PVASASPKLC LAWQGMLLLK NSNFPSNMHL LQGDLQVASS LLVEGSTGGK VAQLKITQRL RLDQPKLDEV TRRIKVAGPN GYAILLAVPG SSDSRSSSSS AASDTATSTQ RPLRNLVSYL KQKQAAGVIS LPVGGNKDKE NTGVLHAFPP CEFSQQFLDS PAKALAKSEE DYLVMIIVRA KLVEQRMKIW NSKL
May function as an mRNA export factor, stimulating export and expression of RTE-containing mRNAs which are present in many retrotransposons that require to be exported prior to splicing. High affinity binding of pre-mRNA to RBM15 may allow targeting of the mRNP to the export helicase DBP5 in a manner that is independent of splicing-mediated NXF1 deposition, resulting in export prior to splicing. May be implicated in HOX gene regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: one twenty two protein; one twenty-two; One-twenty two protein 1; RNA binding motif protein 15; RNA-binding motif protein 15; RNA-binding protein 15
Gene Aliases: OTT; OTT1; RBM15; SPEN
UniProt ID: (Human) Q96T37
Entrez Gene ID: (Human) 64783, (Rat) 684233
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support