Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: GLGKHEQGLS TALSVEKTSK RGGKIIVGDA TEKDASKKSD SNPLTEILKC PTKVVLLRNM VGAGEVDEDL EVETKEECEK YGKVGKCVIF EIPGAPDDEA VRIF
Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
This gene encodes an RNA binding protein. The encoded protein is part of the spliceosome complex and functions in the second catalytic step of mRNA splicing. Alternatively spliced transcript variants have been described. Related pseudogenes exist on chromosomes 9 and 15.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 45 kDa-splicing factor; RNA-binding motif protein 17; Splicing factor 45; splicing factor 45kDa
Gene Aliases: 2700027J02Rik; RBM17; SPF45
UniProt ID: (Human) Q96I25, (Mouse) Q8JZX4
Entrez Gene ID: (Human) 84991, (Rat) 291295, (Mouse) 76938
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support