Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: TQIFVEKLFD AVNTKSYLPP PEQPSSGSLK VEFFPHQEKD IKKEEITKEE EREKKFSRRL NHSPPQSSSR YRENRS; Positive Samples: Mouse brain, Mouse testis, Rat liver
RBM26 (RNA Binding Motif Protein 26) is a Protein Coding gene. Diseases associated with RBM26 include Cutaneous T Cell Lymphoma and Helsmoortel-Van Der Aa Syndrome. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and nucleotide binding. An important paralog of this gene is RBM27. [GeneCards]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cutaneous T-cell lymphoma tumor antigen se70-2; MLZ-393; Protein expressed in male leptotene and zygotene spermatocytes 393; RNA-binding motif protein 26; RNA-binding protein 26; RP11-255E21.1
Gene Aliases: 1700009P03Rik; AI447318; C230097K14Rik; C86225; MLZ-393; Pro1777; Rbm26; RGD1308297; Se70-2
UniProt ID: (Mouse) Q6NZN0
Entrez Gene ID: (Rat) 306137, (Mouse) 74213
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support