Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MTASPDYLVV LFGITAGATG AKLGSDEKEL ILLFWKVVDL ANKKVGQLHE VLVRPDQLEL TEDCKEETKI DVESLSSASQ LDQALRQFNQ SVSNELNIGV GTSFCLCTDG QLHVRQILHP EASKKNVLLP ECFYSFFDLR KEFKKCCPGS PDIDKLDVAT MTEYLNFEKS SSVSRYGASQ VEDMGNIILA MISEPYNHRF SDPERVNYKF ESGTCSKMEL IDDNTVVRAR GLPWQSSDQD IARFFKGLNI AKGGAALCLN AQGRRNGEAL VRFVSEEHRD LALQRHKHHM GTRYIEVYKA TGEDFLKIAG GTSNEVAQFL SKENQVIVRM RGLPFTATAE EVVAFFGQHC PITGGKEGIL FVTYPDGRPT GDAFVLFACE EYAQNALRKH KDLLGKRYIE LFRSTAAEVQ QVLNRFSSAP LIPLPTPPII PVLPQQFVPP TNVRDCIRLR GLPYAATIED ILDFLGEFAT DIRTHGVHMV LNHQGRPSGD AFIQMKSADR AFMAAQKCHK KNMKDRYVEV FQCSAEEMNF VLMGGTLNRN GLSPPPCKLP CLSPPSYTFP APAAVIPTEA AIYQPSVILN PRALQPSTAY YPAGTQLFMN YTAYYPSPPG SPNSLGYFPT AANLSGVPPQ PGTVVRMQGL AYNTGVKEIL NFFQGYQYAT EDGLIHTNDQ ARTLPKEWVC I
RBM35A, also known as ESRP1, is a mRNA splicing factor that with its related protein RBM35B (ESRP2) are coordinators of an epithelial cell-type-specific splicing program. RBM35A contains three putative RNA recognition motifs and acts by directly binding specific sequences in mRNAs. RBM35A is involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5' UTRs of mRNAs. Other recent studies have shown that RMB35A may also act as a novel tumor suppressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epithelial splicing regulatory protein 1; RNA binding motif protein 35A; RNA-binding motif protein 35A; RNA-binding protein 35A
Gene Aliases: ESRP1; RBM35A; RMB35A
UniProt ID: (Human) Q6NXG1
Entrez Gene ID: (Human) 54845
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support