Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish and Bovine
Binds (via the RRM domain) to the 3'-untranslated region (UTR) of CDKN1A mRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC10433; RNA-binding motif protein 42; RNA-binding protein 42
Gene Aliases: 1700003D06Rik; 3100004P22Rik; RBM42
UniProt ID: (Human) Q9BTD8, (Mouse) Q91V81
Entrez Gene ID: (Human) 79171, (Mouse) 68035
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support