Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: IRKMLSSGVC TSTVQLPGKV VVVTGANTGI GKETAKELAQ RGARVYLACR DVEKGELVAK EIQTTTGNQQ VLVRKLDLSD TKSIRAFAKG FLAEEKHLHV LINNAGVMMC PYSKTADGFE MHIGVNHLGH FLLTHLLLEK LKESAPSRIV NVSSLAHHLG RIHFHNLQGE KFYNAGLAYC HSKLANILFT QELARRLKGS GVTTYSVHPG TVQSELVRHS SFMRWMWWLF SFFIKTPQQG AQTSLHCALT EGLEILSGNH FSDCHVAWVS AQARNETIAR RLWDVSCDLL GLPID
RHD11, a member of the short-chain dehydrogenase/reductase superfamily of oxidoreductases, is expressed at high levels in prostate epithelium, and its expression is regulated by androgens.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Androgen-regulated short-chain dehydrogenase/reductase 1; HCV core-binding protein HCBP12; prostate short-chain dehydrogenase reductase 1; Prostate short-chain dehydrogenase/reductase 1; RalR1; Retinal reductase 1; Retinol dehydrogenase 11; Short chain dehydrogenase/reductase family 7C member 1; short chain dehydrogenase/reductase family 7C, member 1
Gene Aliases: ARSDR1; CGI-82; CGI82; HCBP12; MDT1; PSDR1; RALR1; RDH11; RDJCSS; SCALD; SDR7C1
UniProt ID: (Human) Q8TC12
Entrez Gene ID: (Human) 51109
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support