Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLVTLGLLTS FFSFLYMVAP SIRKFFAGGV CRTNVQLPGK VVVITGANTG IGKETARELA SRGARVYIAC RDVLKGESAA SEIRVDTKNS QVLVRKLDLS DTKSIRAFAE GFLAEEKQLH ILINNAGVMM CPYSKTADGF ETHLGVNHLG HFLLTYLLLE QLKVSAPARV VNVSSVAHHI GKIPFHDLQS EKRYSRGFAY CHSKLANVLF TRELAKRLQG TGVTTYAVHP GVVRSELVRH SSLLCLLWRL FSPFVKTARE GAQTSLHCAL AEGLEPLSGK YFSDCKRTWV SPRARNNKTA ERLWNVSCEL LGIRWE
The protein encoded by this gene is an NADPH-dependent retinal reductase whose highest activity is toward 9-cis and all-trans-retinol. The encoded enzyme also plays a role in the metabolism of short-chain aldehydes but does not exhibit steroid dehydrogenase activity. Defects in this gene are a cause of Leber congenital amaurosis type 3 (LCA3).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: All-trans and 9-cis retinol dehydrogenase; LCA3; Retinol dehydrogenase 12; retinol dehydrogenase 12, all-trans and 9-cis; Short chain dehydrogenase/reductase family 7C member 2; short chain dehydrogenase/reductase family 7C, member 2
Gene Aliases: LCA13; RDH12; RP53; SDR7C2
UniProt ID: (Human) Q96NR8
Entrez Gene ID: (Human) 145226
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support