Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
IK Polyclonal Antibody detects endogenous levels of IK protein.
Insulin-like growth factor-II messenger RNA (mRNA)-binding protein-3 (IMP-3), also known as K homology domain-containing protein overexpressed in cancer (KOC) and L523S, is a member of the insulin-like growth factor-II mRNA-binding protein family and is expressed during embryogenesis and in some malignancies. IMP-3 is expressed in malignant melanoma but not in benign nevi, even when dysplastic features are present; IMP-3 is expressed in a significantly higher proportion of melanomas than Spitz nevi; and IMP-3 is expressed in metastatic melanomas significantly more than in thin melanomas. IMP-3 appears to be involved in the progression of malignant melanoma and may play an important role in the regulation of the biologic behavior of this tumor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arginine/glutamic acid/aspartic acid repeat containing protein; chondrosarcoma-associated protein 2; Cytokine IK; Cytokine IK factor; IK factor; prer protein; Protein Red; Protein RER; RD element
Gene Aliases: CSA2; IK; MuRED; RED; RER
UniProt ID: (Human) Q13123, (Rat) Q66HG8, (Mouse) Q9Z1M8
Entrez Gene ID: (Human) 3550, (Rat) 291659, (Mouse) 24010
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support