Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KGYHCTAHK EVGPGPGGGG SGINPAYRTE DANEDTIGVL VRLITEKKEN AAALEELLKE YHSKQLVQTS HRPVSKLPPA PPNVPHICPH RHHLHTVQGL ASLSGPCCSR CSQKKWPEVL LSPEAVAATT PVPSLLPNPT RVPKVGAKAG RQGEITILSV GRFRVARIPE QRTSSMVSEV KTITEAGPSW GDLPDSPQPG LPPEQQALLG SGGSRTKWLK PPAENKAEEN RYVVRLSESN LVI (189-430 aa encoded by BC051810)
The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is especially abundant in hematologic tissues. It has been shown to activate the NF-kappaB pathway and selectively bind TNF receptor-associated factor 1 (TRAF1). This receptor is capable of stimulating T-cell proliferation in the presence of CD3 signaling, which suggests its regulatory role in immune response. Two alternatively spliced transcript variants of this gene encoding the same protein have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Receptor expressed in lymphoid tissues; Tumor necrosis factor receptor superfamily member 19L
Gene Aliases: E430021K24Rik; RELT; TNFRSF19L; TRLT
UniProt ID: (Human) Q969Z4, (Mouse) Q8BX43
Entrez Gene ID: (Human) 84957, (Mouse) 320100, (Rat) 361615
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support