Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Rockland
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This affinity purified antibody is directed against yeast RFA2. This antibody consists of antibodies eluted from the non-phospho peptide column.
Binds to single-stranded sequences participating in DNA replication in addition to those mediating transcriptional repression (URS1) and activation (CAR1). Stimulates the activity of a cognate strand exchange protein (SEP1). It cooperates with T-AG and DNA topoisomerase I to unwind template DNA containing the simian virus 40 origin of DNA replication. [Uniprot]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-binding protein BUF1; Replication factor A protein 2; Replication protein A 36 kDa subunit; RF-A protein 2; Rfa2p
Gene Aliases: BUF1; N0368; RFA2; RPA2; YNL312W
UniProt ID: (Baker's yeast) P26754
Entrez Gene ID: (Baker's yeast) 855404
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support