Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: QPQAQPPGGQ KQYVTELPAV PAPSQPTGAP TPSPAPQQYI VVTVSEGAMR ASETVSEASP GSTASQTGVP TQVVQQVQGT QQRLLVQTSV QAKPGHVSPL QLTNIQVPQQ ALPTQRLVVQ SAAPGSKGGQ VSLTVHGTQQ VHSPPEQSPV QANSSSSKTA GAPTGTVPQQ LQVHGVQQSV PVTQERSVVQ ATPQAPKPGP VQPLTVQGLQ PVHVAQEVQQ LQQVPVPH
This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X2, X3, X4, and X5. It is a transcriptional activator that can bind DNA as a monomer or as a heterodimer with RFX family members X2, X3, and X5, but not with X4. This protein binds to the X-boxes of MHC class II genes and is essential for their expression. Also, it can bind to an inverted repeat that is required for expression of hepatitis B virus genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EF-C; Enhancer factor C; MHC class II regulatory factor RFX; MHC class II regulatory factor RFX1; Regulatory factor X 1; regulatory factor X, 1 (influences HLA class II expression); RFX; trans-acting regulatory factor 1; Transcription factor RFX1; vregulatory factor X, 1 (influences HLA class II expression)
Gene Aliases: AI047719; AI385641; EFC; RFX; RFX1
UniProt ID: (Human) P22670, (Mouse) P48377
Entrez Gene ID: (Human) 5989, (Mouse) 19724
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support