Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQNSEGGADS PASVALRPSA AAPPVPASPQ RVLVQAASSN PKGAQMQPIS LPRVQQVPQQ VQPVQHVYPA QVQYVEGGDA VYTNGAIRTA YTYNPEPQMY APSSTASYFE APGGAQVTVA ASSPPAVPSH SMVGITMDVG GSPIVSSAGA YLIHGGMDST RHSLAHTSRS SPATLEMAIE NLQKSEGITS HKSGLLNSHL QWLLDNYETA EGVSLPRSSL YNHYLRHCQE HKLDPVNAAS FGKLIRSVFM GLRTRRLGTR GNSKYHYYGI RLKPDSPLNR LQEDTQYMAM RQQPMHQKPR YRPAQKTDSL GDSGSHSGLH STPEQTMAVQ SQHHQQYIDV SHVFPEFPAP DLGSFLLQDG VTLHDVKALQ LVYRRHCEAT VDVVMNLQFH YIEKLWLSFW NSKASSSDGP TSLPASDEDP EGAVLPKDKL ISLCQCDPIL RWMRSCDHIL YQALVEILIP DVLRPVPSTL TQAIRNFAKS LEGWLTNAMS DFPQQVIQTK VGVVSAFAQT LRRYTSLNHL AQAARAVLQN TSQINQMLSD LNRVDFANVQ EQASWVCQCE ESVVQRLEQD FKLTLQQQSS LDQWASWLDS VVTQVLKQHA GSPSFPKAAQ QFLLKWSFYS SMVIRDLTLR SAASFGSFHL IRLLYDEYMF YLVEHRVAEA TGETPIAVMG EFNDLASLSL TLLDKDDMGD EQRGSEAGPD ARSLGEPLVK RERSDPNHSL QGI
This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X1, X3, X4, and X5. It is a transcriptional activator that can bind DNA as a monomer or as a heterodimer with other RFX family members. This protein can bind to cis elements in the promoter of the IL-5 receptor alpha gene. Two transcript variants encoding different isoforms have been described for this gene, and both variants utilize alternative polyadenylation sites. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA binding protein RFX2; DNA-binding protein RFX2; FLJ14226; HLA class II regulatory factor RFX2; Regulatory factor X 2; regulatory factor X, 2 (influences HLA class II expression); trans-acting regulatory factor 2
Gene Aliases: RFX2
UniProt ID: (Human) P48378
Entrez Gene ID: (Human) 5990
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support