Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SKSYSYSEVL PIEGPPSFRG QSVKYVYKLT IGCQRVNSPI TLLRVPLRVL VLTGLQDVRF PQDEAVAPSS PFLEEDEGGK KDS
Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Retrograde golgi transport homolog 1 (RGP1) is the mammalian homolog to the yeast RGP1, a protein that forms a tight complex with RIC1. This complex binds Ypt6p and stimulates guanine nucleotide exchange. RGP1 is localized to the Golgi and is thought to be a potential Golgi recycling factor. Rgp1 yeast mutants exhibit defects in retrograde trafficking similar to those seen in yeast with mutations in other retrograde Golgi transport proteins. It is expected that RGP1 plays a similar role in mammalian cells to that seen in yeast.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RAB6A-GEF complex partner protein 2; Retrograde Golgi transport protein RGP1 homolog; RGP1 (HGNC:21965); RGP1 retrograde golgi transport homolog
Gene Aliases: KIAA0258; RGP1
UniProt ID: (Human) Q92546
Entrez Gene ID: (Human) 9827
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support