Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKKHDFAENL PSSLLIIILV IFTLSGSKFG RGTGLTQPSQ RTSARRSFGR SKRFSITRSL DDLESATVSD GELTGADLKD CVSNNSLSSN ASLPSVQSCR RLRERRVASW AVSFERLLQD PVGVRYFSDF LRKEFSEENI LFWQACEYFN HVPAHDKKEL SYRAREIFSK FLCSKATTPV NIDSQAQLAD DVLRAPHPDM FKEQQLQIFN LMKFDSYTRF LKSPLYQECI LAEVEGRALP DSQQVPSSPA SKHSLGSDHS SVSTPKKLSG KSKSGRSLNE ELGDEDSEKK RKGAFFSWSR TRSTGRSQKK REHGDHADDA LHANGGLCRR ESQGSVSSAG SLDLSEACRT LAPEKDKATK HCCIHLPDGT SCVVAVKAGF SIKDILSGLC ERHGINGAAA DLFLVGGDKP LVLHQDSSIL ESRDLRLEKR TLFRLDLVPI NRSVGLKAKP TKPVTEVLRP VVARYGLDLS GLLVRLSGEK EPLDLGAPIS SLDGQRVVLE EKDPSRGKAS ADKQKGVPVK QNTAVNSSSR NHSATSFLSL FPKLRATEQM TNVGC
This gene encodes a member of the 'regulator of G protein signaling' (RGS) gene family. The encoded protein may function as a guanosine triphosphatase (GTPase)-activating protein as well as a transcriptional repressor. This protein may play a role in tumorigenesis. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their biological nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp761K1617; DKFZp761K1817; Regulator of G-protein signaling 12; regulator of G-protein signalling 12; RGS12; RP11-529E10.1
Gene Aliases: RGS12
UniProt ID: (Human) O14924
Entrez Gene ID: (Human) 6002
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support