Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPGKPKHLGV PNGRMVLAVS DGELSSTTGP QGQGEGRGSS LSIHSLPSGP SSPFPTEEQP VASWALSFER LLQDPLGLAY FTEFLKKEFS AENVTFWKAC ERFQQIPASD TQQLAQEARN IYQEFLSSQA LSPVNIDRQA WLGEEVLAEP RPDMFRAQQL QIFNLMKFDS YARFVKSPLY RECLLAEAEG RPLREPGSSR LGSPDATRKK PKLKPGKSLP LGVEELGQLP PVEGPGGRPL RKSFRRELGG TANAALRRES QGSLNSSASL DLGFLAFVSS KSESHRKSLG STEGESESRP GKYCCVYLPD GTASLALARP GLTIRDMLAG ICEKRGLSLP DIKVYLVGNE QALVLDQDCT VLADQEVRLE NRITFELELT ALERVVRISA KPTKRLQEAL QPILEKHGLS PLEVVLHRPG EKQPLDLGKL VSSVAAQRLV LDTLPGVKIS KARDKSPCRS QGCPPRTQDK ATHPPPASPS SLVKVPSSAT GKRQTCDIEG LVELLNRVQS SGAHDQRGLL RKEDLVLPEF LQLPAQGPSS EETPPQTKSA AQPIGGSLNS TTDSAL
RGS14 is a member of the regulator of G-protein signaling family which contains one RGS domain, two Raf-like Ras-binding domains (RBDs), and one GoLoco domain. RGS14 attenuates the signaling activity of G-proteins by binding, through its GoLoco domain, to specific types of activated, GTP-bound G alpha subunits. Acting as a GTPase activating protein (GAP), the protein increases the rate of conversion of the GTP to GDP. The resulting hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal. Alternate transcriptional splice variants of RGS14 have been observed but have not been thoroughly characterized. Further, RGS14 acts as a positive modulator of microtubule polymerisation and spindle organization through a G(i)-alpha-dependent mechanism; plays a role in cell division; is required for the nerve growth factor (NGF)-mediated neurite outgrowth, and is involved in stress resistance. RGS14 protein may also be involved in visual memory processing capacity and hippocampal-based learning and memory.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Regulator of G-protein signaling 14; regulator of G-protein signalling 14; RGS14; RGSE
Gene Aliases: RGS14
UniProt ID: (Human) O43566
Entrez Gene ID: (Human) 10636
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support