Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: METTLLFFSQ INMCESKEKT FFKLIHGSGK EETSKEAKIR AKEKRNRLSL LVQKPEFHED TRSSRSGHLA KETRVSPEEA VKWGESFDKL LSHRDGLEAF TRFLKTEFSE ENIEFWIACE DFKKSKGPQQ IHLKAKAIYE KFIQTDAPKE VNLDFHTKEV ITNSITQPTL HSFDAAQSRV YQLMEQDSYT RFLKSDIYLD LMEGRPQRPT NLRRRSRSFT CNEFQDVQSD VAIWL
This gene encodes a member of the regulator of G-protein signaling family. This protein is contains a conserved, 120 amino acid motif called the RGS domain. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Regulator of G-protein signaling 18; regulator of G-protein signalling 13; regulator of G-protein signalling 18; RGS18; RP11-142L4.1
Gene Aliases: RGS13; RGS18
UniProt ID: (Human) Q9NS28
Entrez Gene ID: (Human) 64407
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support