Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: QQLFQELKG VRLLTDTLEL TLGVTPEGNP PTLLPSQETE RAMEILKVLF NITLDSIKGE VDEEDAALYR HLGTLLRHCV MIATAGDRTE EFHGHAVNLL GNLPLKCLDV LLTLEPHGDS TEFMGVNMDV IRALLIFLEK RLHKTHRLKE SVAPVLSVLT ECARMHRPAR KFLKAQVLPP LRDVRTRPEV GEMLRNKLVR LMTHLDTDVK RVAAEFLFVL CSESVPRFIK YTGYGNAAGL LAARGLMAGG RPEGQYSEDE DTDTDEYKEA KASINPVTGR VEEKPPNPME GMTEEQKEHE AMKLVTMFDK LSRNRVIQPM GMSPRGHLTS LQDAMCETME QQLSSDPDSD PD (117-467 aa encoded by BC011821)
Guanine nucleotide exchange factor (GEF), which can activate some, but not all, G-alpha proteins. Able to activate GNAI1, GNAO1 and GNAQ, but not GNAS by exchanging bound GDP for free GTP. Involved in regulation of microtubule pulling forces during mitotic movement of chromosomes by stimulating G(i)-alpha protein, possibly leading to release G(i)-alpha-GTP and NuMA proteins from the NuMA-GPSM2-G(i)-alpha-GDP complex. Also acts as an activator for G(q)-alpha (GNAQ) protein by enhancing the G(q)-coupled receptor-mediated ERK activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chaperone Ric-8A; heterotrimeric G protein guanine nucleotide exchange factor Ric-8A; MGC104517; MGC131931; MGC148073; MGC148074; resistance to inhibitors of cholinesterase 8 homolog; resistance to inhibitors of cholinesterase 8 homolog A; resistance to inhibitors of cholinesterase 8A; RIC8 protein; synembryn; Synembryn-A
Gene Aliases: AI114950; RIC-8; Ric-8A; RIC8; RIC8A
UniProt ID: (Human) Q9NPQ8, (Mouse) Q3TIR3, (Rat) Q80ZG1
Entrez Gene ID: (Human) 60626, (Mouse) 101489, (Rat) 293614
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support