Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • RIP2 Antibodies

          Abnova

          RIPK2 Polyclonal Antibody, MaxPab™

          View all (23) RIP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite RIPK2 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (3)
          RIPK2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RIPK2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RIPK2 Antibody (H00008767-D01P) in ICC/IF

          Immunofluorescence of purified MaxPab antibody to RIPK2 on HeLa cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RIPK2 Antibody in Immunocytochemistry (ICC/IF)
          RIPK2 Antibody in Western Blot (WB)
          RIPK2 Antibody in Western Blot (WB)

          Product Details

          H00008767-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RIPK2 (NP_003812.1, 1 a.a. approximately 540 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MNGEAICSAL PTIPYHKLAD LRYLSRGASG TVSSARHADW RVQVAVKHLH IHTPLLDSER KDVLREAEIL HKARFSYILP ILGICNEPEF LGIVTEYMPN GSLNELLHRK TEYPDVAWPL RFRILHEIAL GVNYLHNMTP PLLHHDLKTQ NILLDNEFHV KIADFGLSKW RMMSLSQSRS SKSAPEGGTI IYMPPENYEP GQKSRASIKH DIYSYAVITW EVLSRKQPFE DVTNPLQIMY SVSQGHRPVI NEESLPYDIP HRARMISLIE SGWAQNPDER PSFLKCLIEL EPVLRTFEEI TFLEAVIQLK KTKLQSVSSA IHLCDKKKME LSLNIPVNHG PQEESCGSSQ LHENSGSPET SRSLPAPQDN DFLSRKAQDC YFMKLHHCPG NHSWDSTISG SQRAAFCDHK TTPCSSAIIN PLSTAGNSER LQPGIAQQWI QSKREDIVNQ MTEACLNQSL DALLSRDLIM KEDYELVSTK PTRTSKVRQL LDTTDIQGEE FAKVIVQKLK DNKQMGLQPY PEILVVSRSP SLNLLQNKSM

          Target Information

          RIPK2 is a serine/threonine kinase that is involved in cell activation, proliferation, differentiation, and apoptosis. Upon stimulation by bacterial peptidoglycans, NOD1 and NOD2 are activated, oligomerize and recruit RIPK2 through CARD-CARD domains. Contributes to the tyrosine phosphorylation of the guanine exchange factor ARHGEF2 through Src tyrosine kinase leading to NF-kappaB activation by NOD2. Once recruited, RIPK2 autophosphorylates and undergoes 'Lys-63'-linked polyubiquitination by E3 ubiquitin ligases XIAP, BIRC2 and BIRC3. The polyubiquitinated protein mediates the recruitment of MAP3K7/TAK1 to IKBKG/NEMO and induces 'Lys-63'-linked polyubiquitination of IKBKG/NEMO and subsequent activation of IKBKB/IKKB. In turn, NF-kappa-B is released from NF-kappa-B inhibitors and translocates into the nucleus where it activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. Plays also a role during engagement of the T-cell receptor (TCR) in promoting BCL10 phosphorylation and subsequent NF-kappa-B activation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CARD-carrying kinase; CARD-containing IL-1 beta ICE-kinase; CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase; CARD-containing interleukin-1 beta-converting enzyme-associated kinase; EC 2.7.11.1; growth-in; growth-inhibiting gene 30; kinase RIPK2; receptor-interacting protein (RIP)-like interacting caspase-like apoptosis regulatory protein (CLARP) kinase; Receptor-interacting protein 2; receptor-interacting serine-threonine kinase 2; Receptor-interacting serine/threonine-protein kinase 2; RIP 2; RIP-2; RIP-like; RIP-like-interacting CLARP kinase; Tyrosine-protein kinase RIPK2; WUGSC:H_RG437L15.1

          View more View less

          Gene Aliases: CARD3; CARDIAK; CCK; GIG30; RICK; RIP2; RIPK2; UNQ277/PRO314/PRO34092

          View more View less

          UniProt ID: (Human) O43353

          View more View less

          Entrez Gene ID: (Human) 8767

          View more View less

          Function(s)
          protein serine/threonine kinase activity non-membrane spanning protein tyrosine kinase activity signal transducer activity receptor binding protein binding ATP binding LIM domain binding protein homodimerization activity CARD domain binding
          Process(es)
          activation of MAPK activity positive regulation of cytokine-mediated signaling pathway adaptive immune response apoptotic process inflammatory response signal transduction I-kappaB kinase/NF-kappaB signaling JNK cascade positive regulation of peptidyl-threonine phosphorylation peptidyl-tyrosine phosphorylation positive regulation of protein ubiquitination lipopolysaccharide-mediated signaling pathway positive regulation of chemokine production positive regulation of interferon-alpha production positive regulation of interferon-beta production positive regulation of interferon-gamma production positive regulation of interleukin-12 production positive regulation of interleukin-2 production positive regulation of interleukin-6 production positive regulation of tumor necrosis factor production positive regulation of immature T cell proliferation positive regulation of peptidyl-serine phosphorylation toll-like receptor 2 signaling pathway toll-like receptor 4 signaling pathway T cell proliferation positive regulation of apoptotic process negative regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling response to exogenous dsRNA innate immune response positive regulation of T-helper 1 cell differentiation positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade positive regulation of alpha-beta T cell proliferation positive regulation of peptidyl-tyrosine phosphorylation defense response to Gram-positive bacterium T cell receptor signaling pathway positive regulation of NF-kappaB transcription factor activity positive regulation of ERK1 and ERK2 cascade nucleotide-binding oligomerization domain containing signaling pathway nucleotide-binding oligomerization domain containing 1 signaling pathway nucleotide-binding oligomerization domain containing 2 signaling pathway response to interleukin-1 response to interleukin-12 response to interleukin-18 cellular response to lipoteichoic acid cellular response to peptidoglycan cellular response to muramyl dipeptide positive regulation of xenophagy
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0