Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: HLAARNGHLA TVKLLVEEKA DVLARGPLNQ TALHLAAAHG HSEVVEELVS ADVIDLFDEQ GLSALHLAAQ GRHAQTVETL LRHGAHINLQ SLKFQGGHGP AATLLRRSKT
The protein encoded by this gene is a serine/threonine protein kinase that interacts with protein kinase C-delta. The encoded protein can also activate NFkappaB and is required for keratinocyte differentiation. This kinase undergoes autophosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin repeat domain-containing protein 3; PKC-delta-interacting protein kinase; protein kinase C-associated kinase; receptor-interacting serine-threonine kinase 4; serine/threonine-protein kinase ANKRD3
Gene Aliases: ANKK2; ANKRD3; DIK; NKRD3; PKK; PPS2; RIP4
Entrez Gene ID: (Human) 54101
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support