Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KGMTSSQWFK IQHMQPSPQA CNSAMKNINK HTKRCKDLNT FLHEPFSSVA ATCQTPKIAC KNGDKNCHQS HGPVSLTMCK LTSGKYPNCR YKEKRQNKSY VVACKPPQKK DSQQFHLVPV HLDRV
Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 56%.
Binds to an epitope located within the peptide sequence LTMCKLTSGKYPNCR as determined by overlapping synthetic peptides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ribonuclease 7; ribonuclease, RNase A family, 7; RNase 7; SAP-2; Skin-derived antimicrobial protein 2
Gene Aliases: RAE1; RNASE7; UNQ2516/PRO6006
UniProt ID: (Human) Q9H1E1
Entrez Gene ID: (Human) 84659
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support