Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: QPGPSEHSKS FDKWTTKINE CVMKKMNKFY EKRGNFYQNT VGDLLKFIRN LGEHIDEEKH KKMKLKIGDP SLYFQKTFPD LVIYVYTKLQ NTEYRKHFPQ THSPNKPQCD
A virally induced endoribonuclease, RNase L is the terminal factor of the anti-viral action of interferon. Activation of RNase L results in inhibition of viral proliferation. Activation of RNase L has also been determined to degrade RNA, thus, initiating a cellular stress response, inducing apoptosis. Since mutations in RNase L relate to an increased incidence of prostate cancer, the activation of RNase L functions in counteracting prostate cancer, via apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2',5'-oligoisoadenylate synthetase-dependent; 2-5A-dependent ribonuclease; 2-5A-dependent RNase; interferon-induced 2-5A-dependent RNase; Ribonuclease 4; Ribonuclease L; ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent); RNase L
Gene Aliases: PRCA1; RNASEL; RNS4
UniProt ID: (Human) Q05823
Entrez Gene ID: (Human) 6041
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support