Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TDLRTDLSTLM ELSHQKQAPI SYEQGCAIAK QLGAEIYLEG SAFTSEKSIH SI
Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
Lacks intrinsic GTPase activity. Has a low affinity for GDP, and constitutively binds GTP. Controls rearrangements of the actin cytoskeleton. Induces the Rac-dependent neuritic process formation in part by disruption of the cortical actin filaments. Causes the formation of many neuritic processes from the cell body with disruption of the cortical actin filaments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ras homolog gene family, member S; Rho family GTPase 1; Rho-related GTP-binding protein Rho6; Rnd1
Gene Aliases: ARHS; RHO6; RHOS; RND1
UniProt ID: (Human) Q92730
Entrez Gene ID: (Human) 27289
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support