Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKERRASQK LSSKSIMDPN QNVKCKIVVV GDSQCGKTAL LHVFAKDCFP ENYVPTVFEN YTASFEIDTQ RIELSLWDTS GSPYYDNVRP LSYPDSDAVL ICFDISRPET LDSVLKKWKG EIQEFCPNTK MLLVGCKSDL RTDVSTLVEL SNHRQTPVSY DQGANMAKQI GAATYIECSA LQSENSVRDI FHVATLACVN KTNKNVKRNK SQRATKRISH MPSRPELSAV ATDLRKDKAK SCTVM (1-244 aa encoded by BC012513)
Members of the Rho family of Ras-related GTPases, such as ARHE, regulate the organization of the actin cytoskeleton in response to extracellular growth factors. Like Ras, Rho family members appear to cycle between an inactive GDP-bound form and an active GTP-bound form. Three major regulators of Rho activity have been identified: RhoGDIs, which interact with the GDP-bound Rho proteins to keep them in a resting complex; GEFs, which promote GDP/GTP exchange leading to activation of Rho proteins; and GAPs, which stimulate GTP hydrolysis and return the activated Rho protein to its inactive form.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MemB protein; Protein MemB; ras homolog gene family, member E; Rho family GTPase 3; Rho related protein Rnd3/Rho8; Rho-related GTP-binding protein Rho8; Rho-related GTP-binding protein RhoE; rho8; Rnd3; small GTP binding protein Rho8
Gene Aliases: ARHE; memB; RHO8; RHOE; RND3
UniProt ID: (Human) P61587, (Pig) O77683
Entrez Gene ID: (Human) 390, (Pig) 397559
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support