Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: HKTCVDPWLL EHRTCPMCKC DILKALGIEV DVEDGSVSLQ VPVSNEISNS ASSHEEDNRS ETASSGYASV QGTDEPPLEE HVQSTNESLQ LVNHEANSVA VDVIPHVDNP TFEEDETPNQ ETA
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
GRAIL is a type I transmembrane protein that localizes to the endocytic pathway and contains a PA (protease associated) domain and a RING-type zinc finger domain. It is a ubiquitin-protein isopeptide ligase (E3) necessary for the induction of CD4(+) T cell anergy in vivo and induced expression of this protein was observed in anergic CD4 (+) T cells, which suggested a role in the induction of anergic phenotype. It is differentially expressed in naturally occurring and peripherally induced CD25 (+) T regulatory cells and the expression is linked to the conversion of these cells to a regulatory phenotype. Expression of GRAIL in retrovirally transduced T cell hybridomas limits activation-induced IL-2 and IL-4 cytokine production. It also functions in the patterning of the dorsal ectoderm and sensitizes ectoderm to respond to neural-inducing signals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase RNF128; Gene related to anergy in lymphocytes protein; GRAIL; RING finger protein 128; RING-type E3 ubiquitin transferase RNF128; RP11-150F24.1
Gene Aliases: GRAIL; RNF128
UniProt ID: (Human) Q8TEB7
Entrez Gene ID: (Human) 79589
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support