Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen Sequence: LDLMADVVYC PRPCCQLPVM QEPGCTMGIC SSCNFAFCTL CRLTYHGVSP CKVTAEKLMD LRNEYLQADE ANKRLLDQRY GKRVIQKALE EMESKEWLEK N
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein interacts with androgen receptor (AR) and may function as a coactivator that induces AR target gene expression in prostate. A dominant negative mutant of this gene has been demonstrated to inhibit the AR-mediated growth of prostate cancer. This protein also interacts with class III ubiquitin-conjugating enzymes (E2s) and may act as a ubiquitin-ligase (E3) in the ubiquitination of certain nuclear proteins. Five alternatively spliced transcript variants encoding two distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: androgen receptor associated protein 54; Androgen receptor-associated protein 54; E3 ubiquitin-protein ligase RNF14; HFB30; protein Triad2; RING finger protein 14; triad2 protein; TRIAD2 type I
Gene Aliases: 2310075C09Rik; 2610005D23Rik; AA986456; ARA54; AU041447; D18Ertd188e; D7Bwg0165e; HFB30; HRIHFB2038; RNF14; TRIAD2
UniProt ID: (Human) Q9UBS8, (Mouse) Q9JI90
Entrez Gene ID: (Human) 9604, (Rat) 619577, (Mouse) 56736
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support