Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: VSAKSLPNEN FQSLYNKELP VHICNVISPE KIYVQWLLTE NLLNSLEEKM IAAYENSKWE PVKWENDMHC AVKIQDKNQW RRGQIIRMVT DTLVEVLLYD VGVELVVNVD CLRKLEENLK TMGRLSLECS LVDIRPAGGS DKWTATACDC LSLYLTGAVA TIILQVDSEE NNTTWPLPVK IFCRDEKGER VDVSKYLIKK GLALRERRIN NLDNSHSLSE KSLEVPLEQE DSVVTNCIKT NFDPDKKTAD IISEQKVSEF QEKILEPRTT RGYKPPAIPN MNVFEATVSC VGDDGTIFVV P
RNF17 also termed as ring finger protein 17 is a 1623 amino acid protein, which contains 1 ring type zinc finger and 4 tudor domains. RNF17 is specifically expressed in testis. The RING finger motif presents in many ubiquitin E3 ligases. RNF17 interacts with all four members of the Mad family (Mad1, Mxi1, Mad3 and Mad4), which are basic-helix-loop-helix-leucine zipper transcription factors of the Myc oncoprotein network. RNF17 is component of the tectonic-like complex, which is required for ciliogenesis and sonic hedgehog/SHH signaling. RNF17 is able to form dimers or polymers both in vitro and in vivo, indicating that it may play a role in the assembly of RNF17 granules. RNF17 encodes a novel key regulator of spermiogenesis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RING finger protein 17; spermatogenesis associated 23; tudor domain containing 4; Tudor domain-containing protein 4
Gene Aliases: Mmip-2; RNF17; SPATA23; TDRD4
UniProt ID: (Human) Q9BXT8
Entrez Gene ID: (Human) 56163
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support