Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: FYSVSRLPTR SLSDNICAVC GQKIIVELDE EGLIENTYQL SCNHVFHEFC IRGWCIVGKK QTCPYCKEKV DLKRMISNPW ERTHFLYGQI L
RNF175 gene ontology annotations related to this gene include endoplasmic reticulum unfolded protein response; ubiquitin-dependent ERAD pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RING finger protein 175
Gene Aliases: RNF175
UniProt ID: (Human) Q8N4F7
Entrez Gene ID: (Human) 285533
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support