Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: AVTVTVLHP DFTLRLATQA LSQLHARPSY HRLREDVMRL SRLALGSEAW RRVWSRSLQL ASWPNRGGAP GAPQGDPMRV FSVRTRRQDT LPEAGRRSEA EEEEARTIRV TPVRGRERLN EEEPPGGQDP WKLLKEQEER KKCVICQDQS KTVLLLPCRH LCLCQACTEI LMRHPVYHRN CPLCRRGILQ TLNVYL (239-433 aa encoded by BC000058)
The protein encoded by this intronless gene contains a C3HC5 type of RING finger, a motif known to be involved in protein-DNA and protein-protein interactions. The expression of this gene was found to be upregulated in cancer cell lines derived from different types of cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase RNF26; RING finger protein 26; ring finger protein with leucine zipper
Gene Aliases: 8030450I18Rik; RNF26
UniProt ID: (Human) Q9BY78, (Mouse) Q8BUH7
Entrez Gene ID: (Human) 79102, (Mouse) 213211, (Rat) 300659
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support